Assay Details
| Gene Symbol: Dstyk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199463.2 | NP_955750.1 | 9 - 10 | 2302 | 68 | |
| XM_006249774.2 | XP_006249836.1 | 9 - 10 | 2152 | 68 | ||
| GenBank mRNA | AY208851.1 | - | 9 - 10 | 2233 | 68 | |
| AY429675.1 | - | 9 - 10 | 2302 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dstyk
Genomic Map