Assay Details
| Gene Symbol: Slc19a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030024.1 | NP_001025195.1 | 2 - 3 | 963 | 114 | |
| GenBank mRNA | BC099811.1 | - | 2 - 3 | 963 | 7929969 | 114 |
More Information
Updated Mapping Information
Set Membership: |
|

Slc19a2
Genomic Map