Assay Details
| Gene Symbol: Teddm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039915.2 | NP_001035004.1 | 1 | 933 | 104 | |
| GenBank mRNA | AJ515382.1 | - | 1 | 934 | 104 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Teddm1
Genomic Map