Assay Details
| Gene Symbol: Rgl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105957.1 | NP_001099427.1 | 5 - 6 | 807 | 54 | |
| XM_006249982.2 | XP_006250044.1 | 4 - 5 | 921 | 54 | ||
| XM_008769609.1 | XP_008767831.1 | 4 - 5 | 570 | 54 | ||
| XM_008769610.1 | XP_008767832.1 | 3 - 4 | 459 | 54 | ||
| XM_008769611.1 | XP_008767833.1 | 4 - 5 | 570 | 54 | ||
More Information
Updated Mapping Information
Set Membership: | 
		   
					
						
							 | 
				

Rgl1
Genomic Map