Assay Details
Gene Symbol: Chi3l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053560.1 | NP_446012.1 | 1 - 2 | 83 | 103 | |
XM_008769574.1 | XP_008767796.1 | 1 - 2 | 104 | 103 | ||
GenBank mRNA | BC091365.1 | - | 1 - 2 | 83 | 7319171 | 103 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC091365.1,NM_053560.1,XM_008769574.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
