Assay Details
| Gene Symbol: Abl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107186.1 | NP_001100656.1 | 8 - 9 | 1403 | 76 | |
| XM_006250046.2 | XP_006250108.1 | 8 - 9 | 1711 | 76 | ||
| XM_006250047.2 | XP_006250109.1 | 8 - 9 | 1711 | 76 | ||
| XM_006250048.2 | XP_006250110.1 | 8 - 9 | 2382 | 76 | ||
| XM_006250049.2 | XP_006250111.1 | 8 - 9 | 2381 | 76 | ||
| XM_006250050.2 | XP_006250112.1 | 7 - 8 | 1648 | 76 | ||
| XM_006250051.2 | XP_006250113.1 | 7 - 8 | 1648 | 76 | ||
| XM_006250052.2 | XP_006250114.1 | 8 - 9 | 1711 | 76 | ||
| XM_006250053.2 | XP_006250115.1 | 8 - 9 | 1710 | 76 | ||
| XM_006250054.2 | XP_006250116.1 | 8 - 9 | 2380 | 76 | ||
| XM_006250055.2 | XP_006250117.1 | 7 - 8 | 1647 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Abl2
Genomic Map