Assay Details
| Gene Symbol: Srgap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134958.1 | NP_001128430.1 | 9 - 10 | 1854 | 62 | |
| XM_006249782.2 | XP_006249844.1 | 9 - 10 | 1941 | 62 | ||
| XM_006249783.2 | XP_006249845.1 | 9 - 10 | 1825 | 62 | ||
| XM_006249784.2 | XP_006249846.1 | 9 - 10 | 1939 | 62 | ||
| XM_006249785.2 | XP_006249847.1 | 8 - 9 | 2890 | 62 | ||
| XM_008769479.1 | XP_008767701.1 | 9 - 10 | 1936 | 62 | ||
| XM_008769480.1 | XP_008767702.1 | 6 - 7 | 858 | 62 | ||
| XM_008769481.1 | XP_008767703.1 | 6 - 7 | 787 | 62 | ||
| GenBank mRNA | BC168690.1 | - | 9 - 10 | 1854 | 7934193 | 62 |
| FQ212420.1 | - | 2 - 3 | 258 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Srgap2
Genomic Map