Assay Details
| Gene Symbol: Atp1b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013113.2 | NP_037245.2 | 3 - 4 | 516 | 123 | |
| GenBank mRNA | BC078902.1 | - | 3 - 4 | 516 | 7111949 | 123 |
| J02701.1 | - | 3 - 4 | 839 | 123 | ||
| M14137.1 | - | 3 - 4 | 420 | 123 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atp1b1
Genomic Map