Assay Details
| Gene Symbol: Rgs16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077589.1 | NP_001071057.1 | 3 - 4 | 213 | 93 | |
| XM_006250010.2 | XP_006250072.1 | 4 - 5 | 1124 | 93 | ||
| XM_006250011.2 | XP_006250073.1 | 4 - 5 | 1129 | 93 | ||
| XM_006250012.1 | XP_006250074.1 | 4 - 5 | 1190 | 93 | ||
| GenBank mRNA | AY651775.1 | - | 3 - 4 | 213 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rgs16
Genomic Map