Assay Details
| Gene Symbol: Ap3s1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106933.1 | NP_001100403.1 | 1 - 2 | 164 | 102 | |
| XM_006254693.2 | XP_006254755.1 | 1 - 2 | 199 | 102 | ||
| XM_006254694.2 | XP_006254756.1 | 1 - 2 | 197 | 102 | ||
| XM_006254695.2 | XP_006254757.1 | 1 - 2 | 198 | 102 | ||
| XM_006254696.2 | XP_006254758.1 | 1 - 2 | 198 | 102 | ||
| XM_008772131.1 | XP_008770353.1 | 1 - 2 | 198 | 102 | ||
| GenBank mRNA | BC167039.1 | - | 1 - 2 | 146 | 8368273 | 102 |
More Information
Updated Mapping Information
Set Membership: |
|

Ap3s1
Genomic Map