Assay Details
Gene Symbol: Mppe1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108435.2 | NP_001101905.1 | 6 - 7 | 1007 | 60 | |
XM_006254879.2 | XP_006254941.1 | 6 - 7 | 1001 | 60 | ||
XM_006254881.2 | XP_006254943.1 | 6 - 7 | 1653 | 60 | ||
GenBank mRNA | BC162043.1 | - | 6 - 7 | 1007 | 7105580 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
