Assay Details
| Gene Symbol: Cecr5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107884.1 | NP_001101354.1 | 1 - 2 | 141 | 129 | |
| XM_006237246.2 | XP_006237308.1 | 1 - 2 | 604 | 129 | ||
| XM_006237247.2 | XP_006237309.1 | 1 - 2 | 606 | 129 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cecr5
Genomic Map