Assay Details
Gene Symbol: Cecr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107884.1 | NP_001101354.1 | 5 - 6 | 581 | 87 | |
XM_006237246.2 | XP_006237308.1 | 5 - 6 | 1056 | 87 | ||
XM_006237247.2 | XP_006237309.1 | 5 - 6 | 1058 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
