Assay Details
| Gene Symbol: Ccnd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022267.1 | NP_071603.1 | 3 - 4 | 571 | 68 | |
| GenBank mRNA | D16308.1 | - | 3 - 4 | 751 | 68 | |
| FQ227079.1 | - | 3 - 4 | 783 | 68 | ||
| FQ233117.1 | - | 3 - 4 | 783 | 68 | ||
| FQ234359.1 | - | 3 - 4 | 783 | 68 | ||
| L09752.1 | - | 3 - 4 | 571 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ccnd2
Genomic Map