Assay Details
| Gene Symbol: Ccdc77 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191752.1 | NP_001178681.1 | 4 - 5 | 522 | 74 | |
| XM_006237236.2 | XP_006237298.1 | 5 - 6 | 768 | 74 | ||
| XM_006237237.2 | XP_006237299.1 | 5 - 6 | 756 | 74 | ||
| XM_006237238.1 | XP_006237300.1 | 6 - 7 | 811 | 74 | ||
| XM_006237239.1 | XP_006237301.1 | 6 - 7 | 793 | 74 | ||
| XM_006237240.2 | XP_006237302.1 | 5 - 6 | 1679 | 74 | ||
| XM_006237241.1 | XP_006237303.1 | 5 - 6 | 614 | 74 | ||
| XM_006237242.2 | XP_006237304.1 | 4 - 5 | 1699 | 74 | ||
| XM_006237243.2 | XP_006237305.1 | 4 - 5 | 1625 | 74 | ||
| XM_006237244.2 | XP_006237306.1 | 4 - 5 | 522 | 74 | ||
| XM_008763244.1 | XP_008761466.1 | 5 - 6 | 768 | 74 | ||
| XM_008763245.1 | XP_008761467.1 | 5 - 6 | 1753 | 74 | ||
| XM_008763246.1 | XP_008761468.1 | 5 - 6 | 609 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ccdc77
Genomic Map