Assay Details
Gene Symbol: Kcna6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023954.1 | NP_076444.1 | 1 | 3869 | 147 | |
GenBank mRNA | X17621.1 | - | 1 | 2653 | 147 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P384267 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P915100 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 651329 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 438 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 201581 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 659227 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 693795 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 164526 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 15839 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 90307 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 153947 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 627997 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 643762 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 456509 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 474287 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 83969 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 405650 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 890455 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 299798 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 261306 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 707043 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 370713 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 407255 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 887771 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 417411 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 873884 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 577174 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 53577 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 22786 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 140346 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 104040 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 161645 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P384267 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P915100 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P915100 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P384267 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P915100 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P384267 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P16193 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10865739 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10865738 | - | - |
Rat Genome 230 2.0 Array | 1388262_at | - | - |
Rat Genome 230 2.0 Array | 1393124_at | - | - |
Rat Genome 230 2.0 Array | 1388262_PM_at | - | - |
Rat Genome 230 2.0 Array | 1388263_at | - | - |
Rat Genome 230 2.0 Array | 1388263_PM_at | - | - |
Rat Genome 230 2.0 Array | 1393124_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1388263_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1393124_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1388263_at | - | - |
Affymetrix HT Rat Focus Array | 1388262_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1388262_at | - | - |
Affymetrix HT Rat Focus Array | 1393124_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0400026889.rn.1 | TC0400003783.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0400026890.rn.1 | TC0400003783.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0400026885.rn.1 | TC0400003783.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0400026886.rn.1 | TC0400003783.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0400026887.rn.1 | TC0400003783.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0400026888.rn.1 | TC0400003783.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0400026882.rn.1 | TC0400003783.rn.1 | RTA TC |
Rat Expression Set 230 | 1388262_PM_at | - | - |
Rat Expression Set 230 | 1388263_at | - | - |
Rat Expression Set 230 | 1388262_at | - | - |
Rat Expression Set 230 | 1388263_PM_at | - | - |
Rat Expression Set 230 | 1393124_PM_at | - | - |
Rat Expression Set 230 | 1393124_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10865738 | - | - |
Rat Genome U34 Set | M27159cds_s_at | - | - |
Rat Genome U34 Set | X17621cds_at | - | - |
Rat Genome U34 Set | X17621cds_at | - | - |
Rat Genome U34 Set | M27159cds_s_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 20737583 | - | - |
ABI Rat Genome Survey Microarray | 22364986 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1352612 | - | - |
