Assay Details
| Gene Symbol: Gprc5d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109254.1 | NP_001102724.1 | 1 - 2 | 99 | 102 | |
| XM_006237536.2 | XP_006237598.1 | 1 - 2 | 138 | 102 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001109254.1, XM_006237536.2 transcripts.
Set Membership: |
|

Gprc5d
Genomic Map