Assay Details
Gene Symbol: Naaladl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008760899.1 | XP_008759121.1 | 13 - 14 | 3729 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
