Assay Details
Gene Symbol: Lrrc34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044696.1 | NP_001038161.1 | 7 - 8 | 881 | 57 | |
XM_006232235.2 | XP_006232297.1 | 8 - 9 | 852 | 57 | ||
XM_008760937.1 | XP_008759159.1 | 8 - 9 | 943 | 57 | ||
XM_008760938.1 | XP_008759160.1 | 7 - 8 | 639 | 57 | ||
XM_008760939.1 | XP_008759161.1 | 7 - 8 | 835 | 57 | ||
GenBank mRNA | BC097432.1 | - | 7 - 8 | 881 | 7455342 | 57 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
