Assay Details
| Gene Symbol: Skil | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006232237.2 | XP_006232299.1 | 3 - 4 | 2245 | 131 | |
| XM_008760948.1 | XP_008759170.1 | 2 - 3 | 3251 | 131 | ||
| XM_008760949.1 | XP_008759171.1 | 2 - 3 | 3249 | 131 | ||
More Information
Set Membership: |
|

Skil
Genomic Map