Assay Details
| Gene Symbol: Ndufb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106426.1 | NP_001099896.1 | 4 - 5 | 414 | 64 | |
| GenBank mRNA | FQ224246.1 | - | 4 - 5 | 365 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ndufb5
Genomic Map