Assay Details
| Gene Symbol: Depdc1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107651.1 | NP_001101121.1 | 8 - 9 | 1148 | 65 | |
| XM_008760729.1 | XP_008758951.1 | 8 - 9 | 1088 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Depdc1b
Genomic Map