Assay Details
Gene Symbol: Cox7c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134705.1 | NP_001128177.1 | 2 - 3 | 356 | 135 | |
GenBank mRNA | BI289576.1 | - | 1 - 2 | 135 | ||
BQ779631.1 | - | 1 - 2 | 135 | |||
FQ217721.1 | - | 2 - 3 | 305 | 135 | ||
FQ217795.1 | - | 2 - 3 | 305 | 135 | ||
FQ221226.1 | - | 2 - 3 | 366 | 135 | ||
FQ223619.1 | - | 2 - 3 | 307 | 135 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
