Assay Details
| Gene Symbol: Map1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019217.1 | NP_062090.1 | 5 - 6 | 7155 | 62 | |
| XM_008760661.1 | XP_008758883.1 | 4 - 5 | 6820 | 62 | ||
| GenBank mRNA | X16623.1 | - | 1 - 2 | 2370 | 62 | |
| X60370.1 | - | 3 - 4 | 6700 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
microtubule bundle formation
negative regulation of microtubule depolymerization
nervous system development
axonogenesis
synapse assembly
response to wounding
response to mechanical stimulus
response to carbohydrate
cellular process
response to inorganic substance
peripheral nervous system axon regeneration
dendrite development
response to insecticide
developmental maturation
positive regulation of microtubule polymerization
response to nutrient levels
response to estradiol
negative regulation of intracellular transport
response to vitamin A
response to drug
positive regulation of neuron differentiation
positive regulation of axon extension
mitochondrion transport along microtubule
neuron development
axon extension
response to axon injury
induction of synaptic plasticity by chemical substance
establishment of monopolar cell polarity
cellular response to growth factor stimulus
cellular response to peptide hormone stimulus
actin binding
protein binding
phospholipid binding
microtubule binding
protein complex binding
microtubule bundle formation
negative regulation of microtubule depolymerization
nervous system development
axonogenesis
synapse assembly
response to wounding
response to mechanical stimulus
response to carbohydrate
cellular process
response to inorganic substance
peripheral nervous system axon regeneration
dendrite development
response to insecticide
developmental maturation
positive regulation of microtubule polymerization
response to nutrient levels
response to estradiol
negative regulation of intracellular transport
response to vitamin A
response to drug
positive regulation of neuron differentiation
positive regulation of axon extension
mitochondrion transport along microtubule
neuron development
axon extension
response to axon injury
induction of synaptic plasticity by chemical substance
establishment of monopolar cell polarity
cellular response to growth factor stimulus
cellular response to peptide hormone stimulus
actin binding
protein binding
phospholipid binding
microtubule binding
protein complex binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P10887 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 319207 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 383286 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 692701 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 515603 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 377841 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 436557 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 706784 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 558815 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 498699 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 536630 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 861352 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 356447 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 934566 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 764042 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 715019 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 553916 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 489905 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 2688 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 316662 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 946672 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 951408 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 613033 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 58064 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 655002 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 534582 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 164669 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 944387 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 957283 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 611585 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 935307 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 267129 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 569170 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 898071 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 131677 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 467765 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 871026 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 271756 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 621492 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 192899 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 429304 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 280924 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 73222 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 127696 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 938104 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 463730 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 783067 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 360786 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 316271 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 300438 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 356451 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 317306 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 711298 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 158668 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 621805 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 524788 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 55801 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 325617 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 623459 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 561291 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 323114 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 320303 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 245731 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 700712 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 887081 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 585698 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 561548 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 49562 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 650393 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 736476 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 761147 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 704357 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 18099 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 154907 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 18359 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 73414 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 19830 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 163109 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 3498 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 164862 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 32473 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 54633 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 112650 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P10887 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P10887 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P10887 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P182463 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P515029 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P10887 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P1031571 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P287227 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700589641 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 18786 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 5685 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 38898 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 700589641 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10820841 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10820837 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10820840 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10820842 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10820839 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10820836 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10820838 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10820835 | - | - |
| Rat Genome 230 2.0 Array | 1373363_at | - | - |
| Rat Genome 230 2.0 Array | 1395357_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1371003_at | - | - |
| Rat Genome 230 2.0 Array | 1371003_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1395357_at | - | - |
| Rat Genome 230 2.0 Array | 1373363_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1371003_at | - | - |
| Affymetrix HT Rat Focus Array | 1373363_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1395357_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1371003_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1373363_at | - | - |
| Affymetrix HT Rat Focus Array | 1395357_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0200019057.rn.1 | TC0200002718.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0200019058.rn.1 | TC0200002718.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0200019056.rn.1 | TC0200002718.rn.1 | RTA TC |
| Rat Expression Set 230 | 1373363_PM_at | - | - |
| Rat Expression Set 230 | 1371003_at | - | - |
| Rat Expression Set 230 | 1373363_at | - | - |
| Rat Expression Set 230 | 1371003_PM_at | - | - |
| Rat Expression Set 230 | 1395357_PM_at | - | - |
| Rat Expression Set 230 | 1395357_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10820835 | - | - |
| Rat Genome U34 Set | U52950_g_at | - | - |
| Rat Genome U34 Set | U52950_at | - | - |
| Rat Genome U34 Set | X16623cds_s_at | - | - |
| Rat Genome U34 Set | rc_AI008511_at | - | - |
| Rat Genome U34 Set | rc_AI059602_at | - | - |
| Rat Genome U34 Set | rc_AI058961_s_at | - | - |
| Rat Genome U34 Set | rc_AA925503_at | - | - |
| Rat Genome U34 Set | rc_AI229302_s_at | - | - |
| Rat Genome U34 Set | U52950_g_at | - | - |
| Rat Genome U34 Set | X16623cds_s_at | - | - |
| Rat Genome U34 Set | U52950_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22367885 | - | - |
| ABI Rat Genome Survey Microarray | 22398691 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1362692 | - | - |
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_2040245 | - | - |

Map1b
Genomic Map