Assay Details
| Gene Symbol: Bhmt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014256.2 | NP_001014278.2 | 6 - 7 | 799 | 63 | |
| GenBank mRNA | BC079366.1 | - | 6 - 7 | 799 | 7109516 | 63 |
More Information
Updated Mapping Information
Set Membership: |
|

Bhmt2
Genomic Map