Assay Details
Gene Symbol: Marveld2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108936.1 | NP_001102406.1 | 4 - 5 | 1482 | 71 | |
XM_008760679.1 | XP_008758901.1 | 4 - 5 | 1417 | 71 | ||
XM_008760680.1 | XP_008758902.1 | 4 - 5 | 1551 | 71 | ||
XM_008760681.1 | XP_008758903.1 | 4 - 5 | 1519 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
