Assay Details
| Gene Symbol: RGD1560324 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106436.1 | NP_001099906.1 | 2 - 3 | 139 | 122 | |
More Information
Updated Mapping Information
Set Membership: |
|

RGD1560324
Genomic Map