Assay Details
Gene Symbol: Ndufc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003749275.2 | XP_003749323.2 | 2 - 3 | 342 | 150 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
