Assay Details
| Gene Symbol: Frem2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001245978.1 | NP_001232907.1 | 21 - 22 | 8881 | 64 | |
| GenBank mRNA | AB612236.1 | - | 21 - 22 | 8881 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Frem2
Genomic Map