Assay Details
| Gene Symbol: Gmps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024754.1 | NP_001019925.1 | 15 - 16 | 2039 | 111 | |
| GenBank mRNA | BC098016.1 | - | 15 - 16 | 2039 | 7454908 | 111 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P111345 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 61710 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 578053 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 198097 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 154258 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 873483 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 708738 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 322858 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 567839 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 901614 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 341900 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 78396 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 964614 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 312464 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 429395 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 327788 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 437770 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 917504 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 884282 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 315308 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 28521 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 928044 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 172874 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 255194 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 188128 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 854290 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 613907 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 17494 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 71607 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 7488 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 42390 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P111345 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P111345 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P111345 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P18947 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P632078 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 30043 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 62027 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 52813 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10815708 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815712 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815723 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815719 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815707 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815714 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815721 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815711 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815710 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815720 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815716 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815715 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815713 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815709 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815722 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10815706 | - | - |
| Rat Genome 230 2.0 Array | 1380373_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1377577_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1380373_at | - | - |
| Rat Genome 230 2.0 Array | 1377577_at | - | - |
| Affymetrix HT Rat Focus Array | 1377577_at | - | - |
| Affymetrix HT Rat Focus Array | 1380373_at | - | - |
| Affymetrix HT Rat Focus Array | 1380373_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1377577_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0200007949.rn.1 | TC0200001258.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0200007948.rn.1 | TC0200001258.rn.1 | RTA TC |
| Rat Expression Set 230 | 1380373_at | - | - |
| Rat Expression Set 230 | 1380373_PM_at | - | - |
| Rat Expression Set 230 | 1377577_PM_at | - | - |
| Rat Expression Set 230 | 1377577_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815706 | - | - |
| Rat Genome U34 Set | rc_AA999172_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21231240 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1350891 | - | - |

Gmps
Genomic Map