Assay Details
| Gene Symbol: Slc7a11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107673.2 | NP_001101143.2 | 10 - 11 | 1400 | 64 | |
| XM_006232323.2 | XP_006232385.1 | 9 - 10 | 1594 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P576446 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P041166 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 572392 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 105051 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 217426 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 9576 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 806055 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 73734 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 181611 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 471421 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 545087 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 960228 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 455985 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 750644 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 8113 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 632187 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 168685 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 488519 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 338187 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 923732 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 893837 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 576021 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 245442 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 937719 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 412224 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 735128 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 819383 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 208092 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 700158 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 126998 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 303856 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 572231 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 652421 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 715032 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 955660 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 161460 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 664780 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 935494 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 374047 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 57 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 221402 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 875322 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 291561 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 52643 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 726766 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 502527 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 590297 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 477328 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 516809 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 466505 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 943187 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 315059 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 930533 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 657991 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 569738 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 823922 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 715599 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 378091 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 967018 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 47075 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 86060 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 145116 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 69982 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 66888 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 32639 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 36750 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 159974 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 157306 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 117122 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P041166 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P576446 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P041166 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P576446 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P041166 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P576446 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P112201 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 81426 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 81383 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10823069 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10823067 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10823066 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10823058 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10823070 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10823063 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10823065 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10823061 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10823060 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10823064 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10823068 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10823062 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10823059 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10823057 | - | - |
| Rat Genome 230 2.0 Array | 1378133_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1378133_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0200024469.rn.1 | TC0200003626.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0200024476.rn.1 | TC0200003626.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0200024468.rn.1 | TC0200003626.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0200024472.rn.1 | TC0200003626.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0200024466.rn.1 | TC0200003626.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0200024475.rn.1 | TC0200003626.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0200024477.rn.1 | TC0200003626.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0200024474.rn.1 | TC0200003626.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0200024463.rn.1 | TC0200003626.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0200024465.rn.1 | TC0200003626.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0200024471.rn.1 | TC0200003626.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0200024464.rn.1 | TC0200003626.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0200024467.rn.1 | TC0200003626.rn.1 | RTA TC |
| Rat Expression Set 230 | 1378133_PM_at | - | - |
| Rat Expression Set 230 | 1378133_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10823057 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21042242 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1372202 | - | - |

Slc7a11
Genomic Map