Assay Details
| Gene Symbol: Rab25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107687.2 | NP_001101157.2 | 2 - 3 | 466 | 106 | |
| GenBank mRNA | BC168745.1 | - | 2 - 3 | 463 | 6920264 | 106 |
More Information
Updated Mapping Information
Set Membership: |
|

Rab25
Genomic Map