Assay Details
| Gene Symbol: Rab25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107687.2 | NP_001101157.2 | 1 - 2 | 265 | 110 | |
| GenBank mRNA | BC168745.1 | - | 1 - 2 | 262 | 6920264 | 110 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC168745.1, NM_001107687.2 transcripts.
Set Membership: |
|

Rab25
Genomic Map