Assay Details
Gene Symbol: Rxfp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201417.1 | NP_958820.1 | 17 - 18 | 1968 | 116 | |
XM_006232508.2 | XP_006232570.1 | 17 - 18 | 2105 | 116 | ||
XM_006232510.2 | XP_006232572.1 | 14 - 15 | 1659 | 116 | ||
XM_008761085.1 | XP_008759307.1 | 13 - 14 | 3409 | 116 | ||
XM_008761086.1 | XP_008759308.1 | 14 - 15 | 2449 | 116 | ||
XM_008761087.1 | XP_008759309.1 | 13 - 14 | 1591 | 116 | ||
XM_008761088.1 | XP_008759310.1 | 13 - 14 | 1523 | 116 | ||
GenBank mRNA | AY509976.1 | - | 17 - 18 | 1968 | 116 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
adenylate cyclase-modulating G-protein coupled receptor signaling pathway
adenylate cyclase-activating G-protein coupled receptor signaling pathway
parturition
cell differentiation
extracellular matrix organization
lung connective tissue development
nipple morphogenesis
G-protein coupled receptor activity
hormone binding
metal ion binding
adenylate cyclase-modulating G-protein coupled receptor signaling pathway
adenylate cyclase-activating G-protein coupled receptor signaling pathway
parturition
cell differentiation
extracellular matrix organization
lung connective tissue development
nipple morphogenesis
G-protein coupled receptor activity
hormone binding
metal ion binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P142620 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 69774 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 368231 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 623888 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 565174 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 961699 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 244662 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 173855 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 187347 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 877536 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 952041 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 953051 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 58634 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 557567 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 736344 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 470219 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 627684 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 956453 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 249740 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 261158 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 438294 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 270337 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 744619 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 915262 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 693986 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 758991 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 331369 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 474270 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 758245 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 57841 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 93080 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 366441 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 49944 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 167566 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 592296 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 779932 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 747264 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 515172 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 491620 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 451176 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 369113 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 430685 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 460873 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 497251 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 643538 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 923594 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 555097 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 967828 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 558709 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 159948 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 759246 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 150359 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 241742 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 298716 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 526692 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 202930 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 353757 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 375251 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 583968 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 372357 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 696898 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 163735 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 57642 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 275530 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 751924 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 164853 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 196613 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 469663 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 342441 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 937172 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 465462 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 759649 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 391173 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 243878 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 238651 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 646881 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 444861 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 192100 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 665321 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 107086 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 351442 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 371930 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 427896 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 517009 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 802852 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 118401 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 40808 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 471304 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 41654 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 137360 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 170724 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 79864 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 134678 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 73566 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 159290 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 130971 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 153213 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 22286 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 60282 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 92899 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 169364 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 32160 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 104210 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P142620 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P142620 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P142620 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P287908 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 15365 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 52182 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 2132 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 46267 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 52561 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 98770 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10823821 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10823823 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10823831 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10823833 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10823825 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10823836 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10823820 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10823835 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10823830 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10823829 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10823832 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10823826 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10823834 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10823822 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10823828 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10823824 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10823837 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10823827 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10823819 | - | - |
Affymetrix Rat Clariom S Assay | PSR0200026257.rn.1 | TC0200003913.rn.1 | RTA TC |
Affymetrix Rat Gene 1.0 ST Array | 10823819 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22022476 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1348992 | - | - |
RatRef-12 Expression BeadChip | ILMN_49664 | - | - |
