Assay Details
Gene Symbol: Lgals3bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139096.1 | NP_620796.1 | 4 - 5 | 529 | 74 | |
GenBank mRNA | AF065438.1 | - | 4 - 5 | 529 | 74 | |
AY552591.1 | - | 3 - 4 | 391 | 74 | ||
BC081724.1 | - | 4 - 5 | 502 | 7123606 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
