Assay Details
Gene Symbol: Srebf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276707.1 | NP_001263636.1 | 5 - 6 | 1126 | 79 | |
NM_001276708.1 | NP_001263637.1 | 5 - 6 | 1013 | 79 | ||
XM_008767879.1 | XP_008766101.1 | 5 - 6 | 1266 | 79 | ||
GenBank mRNA | AF286469.2 | - | 5 - 6 | 980 | 79 | |
AF286470.2 | - | 5 - 6 | 980 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
