Assay Details
| Gene Symbol: Mrpl12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029900.1 | NP_001025071.1 | 1 - 2 | 302 | 147 | |
| GenBank mRNA | FQ228179.1 | - | 1 - 2 | 116 | 147 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mrpl12
Genomic Map