Assay Details
| Gene Symbol: Mrpl12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029900.1 | NP_001025071.1 | 2 - 3 | 485 | 80 | |
| GenBank mRNA | FQ228179.1 | - | 2 - 3 | 299 | 80 | |
| FQ233456.1 | - | 2 - 3 | 301 | 80 | ||
| FQ234892.1 | - | 2 - 3 | 299 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mrpl12
Genomic Map