Assay Details
| Gene Symbol: Vdac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031353.1 | NP_112643.1 | 5 - 6 | 384 | 83 | |
| XM_006246346.1 | XP_006246408.1 | 4 - 5 | 357 | 83 | ||
| GenBank mRNA | AB039662.1 | - | 5 - 6 | 384 | 83 | |
| AF048828.1 | - | 5 - 6 | 374 | 83 | ||
| AF268467.1 | - | 4 - 5 | 318 | 83 | ||
| BC060558.1 | - | 5 - 6 | 361 | 83 | ||
| BC072484.1 | - | 5 - 6 | 368 | 7105146 | 83 | |
| BC087573.1 | - | 5 - 6 | 363 | 7304939 | 83 | |
| BC087657.1 | - | 5 - 6 | 340 | 7312189 | 83 | |
| BC104684.1 | - | 5 - 6 | 354 | 7462139 | 83 | |
| BC127491.1 | - | 5 - 6 | 340 | 8370617 | 83 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Vdac1
Genomic Map