Assay Details
Gene Symbol: Vdac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031353.1 | NP_112643.1 | 6 - 7 | 620 | 97 | |
XM_006246346.1 | XP_006246408.1 | 5 - 6 | 593 | 97 | ||
GenBank mRNA | AB039662.1 | - | 6 - 7 | 620 | 97 | |
AF048828.1 | - | 6 - 7 | 664 | 97 | ||
AF268467.1 | - | 5 - 6 | 554 | 97 | ||
BC060558.1 | - | 6 - 7 | 597 | 97 | ||
BC072484.1 | - | 6 - 7 | 604 | 7105146 | 97 | |
BC087573.1 | - | 6 - 7 | 599 | 7304939 | 97 | |
BC087657.1 | - | 6 - 7 | 576 | 7312189 | 97 | |
BC104684.1 | - | 6 - 7 | 590 | 7462139 | 97 | |
BC127491.1 | - | 6 - 7 | 576 | 8370617 | 97 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
