Assay Details
Gene Symbol: Fam64a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113781.2 | NP_001107253.2 | 2 - 3 | 334 | 73 | |
XM_006246759.2 | XP_006246821.1 | 2 - 3 | 444 | 73 | ||
XM_006246760.2 | XP_006246822.1 | 2 - 3 | 420 | 73 | ||
GenBank mRNA | BC158692.1 | - | 2 - 3 | 334 | 9022735 | 73 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
