Assay Details
Gene Symbol: Med11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105799.1 | NP_001099269.1 | 1 - 2 | 143 | 91 | |
XM_006246660.2 | XP_006246722.1 | 1 - 2 | 481 | 91 | ||
GenBank mRNA | FQ221563.1 | - | 1 - 2 | 155 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
