Assay Details
| Gene Symbol: Ace3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258237.1 | NP_001245166.1 | 3 - 4 | 391 | 88 | |
| GenBank mRNA | BC085122.1 | - | 3 - 4 | 391 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ace3
Genomic Map