Assay Details
Gene Symbol: Arsg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047877.1 | NP_001041342.1 | 5 - 6 | 708 | 64 | |
XM_006247599.2 | XP_006247661.1 | 7 - 8 | 2908 | 64 | ||
XM_006247600.2 | XP_006247662.1 | 6 - 7 | 2765 | 64 | ||
XM_006247601.2 | XP_006247663.1 | 8 - 9 | 3134 | 64 | ||
XM_006247602.2 | XP_006247664.1 | 7 - 8 | 1696 | 64 | ||
XM_006247603.2 | XP_006247665.1 | 7 - 8 | 1550 | 64 | ||
XM_006247604.2 | XP_006247666.1 | 7 - 8 | 2966 | 64 | ||
XM_006247606.2 | XP_006247668.1 | 6 - 7 | 2823 | 64 | ||
XM_006247607.2 | XP_006247669.1 | 6 - 7 | 1735 | 64 | ||
XM_006247608.2 | XP_006247670.1 | 5 - 6 | 1385 | 64 | ||
XM_006247609.2 | XP_006247671.1 | 5 - 6 | 680 | 64 | ||
XM_008768368.1 | XP_008766590.1 | 8 - 9 | 3077 | 64 | ||
XM_008768369.1 | XP_008766591.1 | 9 - 10 | 2541 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
