Assay Details
| Gene Symbol: Plcd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105845.2 | NP_001099315.2 | 10 - 11 | 1793 | 60 | |
| XM_006247494.2 | XP_006247556.1 | 10 - 11 | 1834 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Plcd3
Genomic Map