Assay Details
| Gene Symbol: Pgs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006247855.2 | XP_006247917.1 | 2 - 3 | 352 | 83 | |
| XM_221142.8 | XP_221142.5 | 2 - 3 | 352 | 83 | ||
More Information
Set Membership: |
|

Pgs1
Genomic Map