Assay Details
| Gene Symbol: Cenpi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012955.1 | NP_037087.1 | 13 - 14 | 1479 | 66 | |
| XM_006257215.2 | XP_006257277.1 | 13 - 14 | 1617 | 66 | ||
| XM_006257216.2 | XP_006257278.1 | 13 - 14 | 1778 | 66 | ||
| XM_006257217.2 | XP_006257279.1 | 12 - 13 | 1492 | 66 | ||
| XM_006257218.1 | XP_006257280.1 | 11 - 12 | 1144 | 66 | ||
| XM_008773403.1 | XP_008771625.1 | 12 - 13 | 1833 | 66 | ||
| GenBank mRNA | X90355.1 | - | 13 - 14 | 1479 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cenpi
Genomic Map