Assay Details
Gene Symbol: Abcb7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212518.1 | NP_997683.1 | 15 - 16 | 2055 | 92 | |
GenBank mRNA | AJ621255.1 | - | 15 - 16 | 2055 | 92 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P21510 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 250109 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 838470 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 177961 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 284026 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 486840 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 929630 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 95338 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 695782 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 677278 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 260516 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 413568 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 144763 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 922411 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 186592 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 553528 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 191966 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 329981 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 394584 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 668273 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 733288 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 569626 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 164208 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 442081 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 382858 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 226611 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 839941 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 711604 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 881694 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 245510 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 618748 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 473002 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 474878 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 945710 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 797284 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 213985 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 305824 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 708528 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 31924 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 600253 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 168688 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 541026 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 104810 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 777484 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 116439 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 600037 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 664717 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 899124 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 409914 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 919383 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 570144 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 616885 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 207321 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 725805 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 794132 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 895210 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 308911 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 729764 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 489769 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 948984 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 392777 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 579506 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 68206 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 896257 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 945251 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 497470 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 95229 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 965776 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 893892 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 442306 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 96480 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 335821 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 15205 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 242989 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 839888 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 336310 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 877110 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 193791 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 677286 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 208906 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 34063 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 778864 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 82615 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 77540 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 324273 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 380622 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 179967 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 825964 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 2751 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 5429 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 19496 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 160928 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 102674 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 131550 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 162060 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 90200 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 140646 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 36559 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 80381 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 91386 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 17348 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 161937 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 21873 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P21510 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P21510 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P21510 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P21510 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 3010 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 65142 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 22455 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 22781 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 44582 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 18776 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 78333 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 64137 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 50858 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 72744 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10938870 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938872 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938863 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938859 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938866 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938873 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938862 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938860 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938869 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938867 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938865 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938871 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938864 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938861 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938868 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10938858 | - | - |
Affymetrix Rat Clariom S Assay | PSR0X00012795.rn.1 | TC0X00002025.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0X00012797.rn.1 | TC0X00002025.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0X00012794.rn.1 | TC0X00002025.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0X00012796.rn.1 | TC0X00002025.rn.1 | RTA TC |
Affymetrix Rat Gene 1.0 ST Array | 10938858 | - | - |
Rat Genome U34 Set | rc_AI072132_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21847687 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1361471 | - | - |
RatRef-12 Expression BeadChip | ILMN_61124 | - | - |
