Assay Details
Gene Symbol: Rpl36a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128065.1 | NP_001121537.1 | 1 - 2 | 51 | 66 | |
GenBank mRNA | FQ211253.1 | - | 1 - 2 | 54 | 66 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the NM_001128065.1 transcripts.
This assay is in the 5'UTR region of the NM_001128065.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
