Assay Details
Gene Symbol: Gas5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_002704.1 | 4 - 5 | 139 | 73 | ||
GenBank mRNA | U77829.1 | - | 4 - 5 | 139 | 73 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
